accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 568577-83-3

CH3SNFFFSOOCl

CAS RN 568577-83-3

IUPAC Name: 4-methyl-2-[4-(trifluoromethyl)phenyl]-1,3-thiazole-5-sulfonyl chloride
Synonyms: 4-methyl-2-[4-(trifluoromethyl)phenyl]-1,3-thiazole-5-sulfonyl chloride
Molecular Weight (g/mol): 341.75
Molecular Formula: C11H7ClF3NO2S2
InChi Key: FIPBWMJINRBLKG-UHFFFAOYSA-N
SMILES: CC1=C(SC(=N1)C1=CC=C(C=C1)C(F)(F)F)S(Cl)(=O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results