accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 568577-88-8

IUPAC Name: 4-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]morpholine
Synonyms: 4-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]morpholine
Molecular Weight (g/mol): 289.18
Molecular Formula: C16H24BNO3
InChi Key: UCPALIMHMYIZPZ-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC=C(C=C1)N1CCOCC1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results