CAS RN 56879-46-0
CAS RN 56879-46-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(+/-)-Homoproline, TRC
CAS: 56879-46-0 Molecular Formula: C6 H11 N O2 Molecular Weight (g/mol): 129.16 Synonym: 2-Pyrrolidineacetic acid,(±)-Homoproline,2-(Pyrrolidin-2-yl)acetic acid IUPAC Name: 2-pyrrolidin-2-ylacetic acid SMILES: OC(=O)CC1CCCN1