accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 56881-31-3

NNSOHOH2N

CAS RN 56881-31-3

IUPAC Name: 7-aminothieno[2,3-b]pyrazine-6-carboxylic acid
Synonyms: 7-aminothieno[2,3-b]pyrazine-6-carboxylic acid
Molecular Weight (g/mol): 195.2
Molecular Formula: C7H5N3O2S
InChi Key: UJUCBOIXAMPUQL-UHFFFAOYSA-N
SMILES: NC1=C(SC2=NC=CN=C12)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results