accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 56891-59-9

IUPAC Name: N-(4-amino-2-methylphenyl)acetamide
Synonyms: N-(4-amino-2-methylphenyl)acetamide
Molecular Weight (g/mol): 164.21
Molecular Formula: C9H12N2O
InChi Key: GWFPMSIIVJMYRZ-UHFFFAOYSA-N
SMILES: CC(=O)NC1=CC=C(N)C=C1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results