accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 56908-88-4

BrBrBr

CAS RN 56908-88-4

IUPAC Name: 1,3-dibromo-5-(bromomethyl)benzene
Synonyms: 1,3-dibromo-5-(bromomethyl)benzene
Molecular Weight (g/mol): 328.83
Molecular Formula: C7H5Br3
InChi Key: PWTFRUXTAFBWBW-UHFFFAOYSA-N
SMILES: BrCC1=CC(Br)=CC(Br)=C1
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results