accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 56961-25-2

IUPAC Name: 4-amino-3,5-dichlorobenzoic acid
Synonyms: 4-amino-3,5-dichlorobenzoic acid
Molecular Weight (g/mol): 206.02
Molecular Formula: C7H5Cl2NO2
InChi Key: UHXYYTSWBYTDPD-UHFFFAOYSA-N
SMILES: NC1=C(Cl)C=C(C=C1Cl)C(O)=O
Percent Purity 
  • (1)
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results