accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3BrOHO

CAS RN 56970-78-6

IUPAC Name: 3-bromo-2-methylpropanoic acid
Synonyms: 3-bromo-2-methylpropanoic acid
Molecular Weight (g/mol): 167
Molecular Formula: C4H7BrO2
InChi Key: BUPXDXGYFXDDAA-UHFFFAOYNA-N
SMILES: CC(CBr)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results