accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 57-00-1

IUPAC Name: 2-(N-methylcarbamimidamido)acetic acid
Synonyms: creatine krebiozon kreatin pyrolysate methylguanidoacetate methylglycocyamine methylguanidoacetic acid cosmocair C 100 N-methyl-N-guanylglycine phosphagen
Molecular Weight (g/mol): 131.14
Molecular Formula: C4H9N3O2
InChi Key: CVSVTCORWBXHQV-UHFFFAOYSA-N
SMILES: CN(CC(O)=O)C(N)=N
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results