accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 57-11-4

IUPAC Name: octadecanoic acid
Synonyms: stearic acid acide stearique hydrofol acid 1855 hydrofol acid 1655 industrene 5016 stearic acid, ion(1-) (8CI) glycon TP glycon DP acidum stearinicul hydrofol acid 150
Molecular Weight (g/mol): 284.48
Molecular Formula: C18H36O2
InChi Key: QIQXTHQIDYTFRH-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCC(O)=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)
  • (3)
  • (1)
Boiling Point 
  • (1)
  • (4)
  • (2)
  • (3)
Physical Form 
  • (1)
  • (3)
  • (2)
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (4)
  • (2)
  • (3)
  • (1)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)