accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 57-67-0

IUPAC Name: N''-(4-aminobenzenesulfonyl)guanidine
Synonyms: sulfaguanidine 磺胺脒
Molecular Weight (g/mol): 214.24
Molecular Formula: C7H10N4O2S
InChi Key: BRBKOPJOKNSWSG-UHFFFAOYSA-N
SMILES: NC(N)=NS(=O)(=O)C1=CC=C(N)C=C1
Molecular Weight (g/mol) 
  • (6)
  • (3)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Certifications/Compliance 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (6)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)