accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3CH3NNHSOOH2NN

CAS RN 57-68-1

IUPAC Name: 4-amino-N-(4,6-dimethylpyrimidin-2-yl)benzene-1-sulfonamide
Synonyms: sulfamethazine sulfametazyny kelametazine intradine sulfadimezinum hava-span sulfametazina 磺胺二甲嘧啶 sulfadimethyldiazine sulphamidine
Molecular Weight (g/mol): 278.33
Molecular Formula: C12H14N4O2S
InChi Key: ASWVTGNCAZCNNR-UHFFFAOYSA-N
SMILES: CC1=CC(C)=NC(NS(=O)(=O)C2=CC=C(N)C=C2)=N1
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
Certifications/Compliance 
  • (3)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)