accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CO(S)(S)(S)O(R)H3C(S)(S)H3C

CAS RN 57-83-0

IUPAC Name: (1S,3aS,3bS,9aR,9bS,11aS)-1-acetyl-9a,11a-dimethyl-1H,2H,3H,3aH,3bH,4H,5H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-one
Synonyms: progesterone proluton glanducorpin prolutone prolutin akrolutin lutocylin lutocylol progesterol pranone
Molecular Weight (g/mol): 314.47
Molecular Formula: C21H30O2
InChi Key: RJKFOVLPORLFTN-LEKSSAKUSA-N
SMILES: CC(=O)[C@H]1CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Industry Type 
  • (1)
  • (1)
  • (3)
Certifications/Compliance 
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (2)
  • (1)
  • (3)
  • (1)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (1)
  • (3)
  • (2)
  • (1)