accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 570-24-1

CH3H2NNOO

CAS RN 570-24-1

IUPAC Name: 2-methyl-6-nitroaniline
Synonyms: 6-nitro-o-toluidine
Molecular Weight (g/mol): 152.15
Molecular Formula: C7H8N2O2
InChi Key: FCMRHMPITHLLLA-UHFFFAOYSA-N
SMILES: CC1=CC=CC(=C1N)[N+]([O-])=O
Molecular Weight (g/mol) 
  • (3)
  • (4)
Quantity 
  • (1)
  • (2)
  • (3)
  • (1)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (3)
  • (4)
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)