accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 5707-04-0

ClSe

CAS RN 5707-04-0

IUPAC Name: (phenylselanyl)chlorane
Synonyms: (phenylselanyl)chlorane
Molecular Weight (g/mol): 191.53
Molecular Formula: C6H5ClSe
InChi Key: WJCXADMLESSGRI-UHFFFAOYSA-N
SMILES: Cl[Se]C1=CC=CC=C1
Boiling Point 
  • (2)
  • (4)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (4)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (4)