accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3H3CCH3OONNNNH2

CAS RN 571188-59-5

IUPAC Name: tert-butyl 4-(6-aminopyridin-3-yl)piperazine-1-carboxylate
Synonyms: tert-butyl 4-(6-aminopyridin-3-yl)piperazine-1-carboxylate
Molecular Weight (g/mol): 278.36
Molecular Formula: C14H22N4O2
InChi Key: RMULRXHUNOVPEI-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1CCN(CC1)C1=CN=C(N)C=C1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results