accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HHClClH2NNH2

CAS RN 57166-92-4

IUPAC Name: dihydrogen methanediamine dichloride
Synonyms: dihydrogen methanediamine dichloride
Molecular Weight (g/mol): 118.99
Molecular Formula: CH8Cl2N2
InChi Key: QCYJCJJCNRIMNG-UHFFFAOYSA-N
SMILES: [H+].[H+].[Cl-].[Cl-].NCN
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results