accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OOCH3P

CAS RN 5717-37-3

IUPAC Name: ethyl 2-(triphenyl-λ⁵-phosphanylidene)propanoate
Synonyms: ethyl 2-(triphenyl-λ⁵-phosphanylidene)propanoate
Molecular Weight (g/mol): 362.41
Molecular Formula: C23H23O2P
InChi Key: KZENFXVDPUMQOE-UHFFFAOYSA-N
SMILES: CCOC(=O)C(C)=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (6)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (4)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (4)
  • (1)
  • (1)
  • (6)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (4)
  • (2)
  • (2)