accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 572-09-8

(S)(R)(R)(R)O(R)OOOOOOOOBrH3CCH3CH3CH3

CAS RN 572-09-8

IUPAC Name: [(2R,3R,4S,5R,6R)-3,4,5-tris(acetyloxy)-6-bromooxan-2-yl]methyl acetate
Synonyms: [(2R,3R,4S,5R,6R)-3,4,5-tris(acetyloxy)-6-bromooxan-2-yl]methyl acetate
Molecular Weight (g/mol): 411.2
Molecular Formula: C14H19BrO9
InChi Key: CYAYKKUWALRRPA-RGDJUOJXSA-N
SMILES: CC(=O)OC[C@H]1O[C@H](Br)[C@H](OC(C)=O)[C@@H](OC(C)=O)[C@@H]1OC(C)=O
Molecular Weight (g/mol) 
  • (3)
  • (4)
Quantity 
  • (3)
  • (1)
  • (3)
Percent Purity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (3)
  • (3)
  • (4)
  • (3)
  • (1)
  • (3)
  • (2)
  • (2)
  • (2)