CAS RN 572-83-8
IUPAC Name:
2-bromo-9,10-dihydroanthracene-9,10-dione
Synonyms:
2-bromoanthracene-9,10-dione
Molecular Weight (g/mol):
287.11
Molecular Formula:
C14H7BrO2
InChi Key:
VTSDGYDTWADUJQ-UHFFFAOYSA-N
SMILES:
BrC1=CC=C2C(=O)C3=CC=CC=C3C(=O)C2=C1
1
–
1
of
1
results
1
2-Bromoanthraquinone, Thermo Scientific Chemicals
CAS: 572-83-8 Molecular Formula: C14H7BrO2 Molecular Weight (g/mol): 287.11 InChI Key: VTSDGYDTWADUJQ-UHFFFAOYSA-N IUPAC Name: 2-bromo-9,10-dihydroanthracene-9,10-dione SMILES: BrC1=CC=C2C(=O)C3=CC=CC=C3C(=O)C2=C1