accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NH3C

CAS RN 5724-56-1

IUPAC Name: 2,3-dimethylbenzonitrile
Synonyms: benzonitrile, 2,3-dimethyl-
Molecular Weight (g/mol): 131.18
Molecular Formula: C9H9N
InChi Key: ZEDPQIJYJCPIRM-UHFFFAOYSA-N
SMILES: CC1=CC=CC(C#N)=C1C
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)