accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3H3CCH3OONN

CAS RN 57260-70-5

IUPAC Name: tert-butyl 4-benzylpiperazine-1-carboxylate
Synonyms: tert-butyl 4-benzylpiperazine-1-carboxylate
Molecular Weight (g/mol): 276.38
Molecular Formula: C16H24N2O2
InChi Key: GVHSMUYEAWMYLM-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1CCN(CC2=CC=CC=C2)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results