CAS RN 57383-80-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2,3-Dibromophenol, TRC
CAS: 57383-80-9 Molecular Formula: C6H4Br2O Molecular Weight (g/mol): 251.9 Synonym: Phenol, 2,3-dibromo-,2,3-Dibromophenol SMILES: Oc1cccc(Br)c1Br