accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 57395-89-8

IUPAC Name: 4-fluoropiperidine hydrochloride
Synonyms: 4-fluoropiperidine hydrochloride
Molecular Weight (g/mol): 139.6
Molecular Formula: C5H11ClFN
InChi Key: IXENWFQXVCOHAZ-UHFFFAOYSA-N
SMILES: Cl.FC1CCNCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results