accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 574-17-4

IUPAC Name: 1-acetyl-2,3-dihydro-1H-indole-2,3-dione
Synonyms: acetylisatin
Molecular Weight (g/mol): 189.17
Molecular Formula: C10H7NO3
InChi Key: LPGDEHBASRKTDG-UHFFFAOYSA-N
SMILES: CC(=O)N1C(=O)C(=O)C2=CC=CC=C12
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results