accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHO

CAS RN 574-96-9

IUPAC Name: 1-hydroxynaphthalene-2-carbaldehyde
Synonyms: 1-hydroxy-2-naphthaldehyde
Molecular Weight (g/mol): 172.18
Molecular Formula: C11H8O2
InChi Key: OITQDWKMIPXGFL-UHFFFAOYSA-N
SMILES: OC1=C2C=CC=CC2=CC=C1C=O
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)