accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NNCH3HOO

CAS RN 5744-56-9

IUPAC Name: 1,3-dimethyl-1H-pyrazole-5-carboxylic acid
Synonyms: 2,5-dimethylpyrazole-3-carboxylic acid
Molecular Weight (g/mol): 140.14
Molecular Formula: C6H8N2O2
InChi Key: QRWZFUBHOQWUGH-UHFFFAOYSA-N
SMILES: CN1N=C(C)C=C1C(O)=O
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results