CAS RN 57452-97-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(S)-Praziquantel, TRC
CAS: 57452-97-8 Molecular Formula: C19 H24 N2 O2 Molecular Weight (g/mol): 312.41 Synonym: (11bS)-2-(Cyclohexylcarbonyl)-1,2,3,6,7,11b-hexahydro-4H-pyrazino[2,1-a]isoquinolin-4-one,(S)-2-(Cyclohexylcarbonyl)-1,2,3,6,7,11b-hexahydro-4H-pyrazino[2,1-a]isoquinolin-4-one,(+)-Praziquantel,(S)-(+)-Praziquantel,(S)-Praziquantel,d-Praziquantel IUPAC Name: (11bS)-2-(cyclohexanecarbonyl)-3,6,7,11b-tetrahydro-1H-pyrazino[2,1-a]isoquinolin-4-one SMILES: O=C(C1CCCCC1)N2C[C@H]3N(CCc4ccccc34)C(=O)C2