accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OOCl

CAS RN 57489-93-7

IUPAC Name: 5-phenylfuran-2-carbonyl chloride
Synonyms: 5-phenylfuran-2-carbonyl chloride
Molecular Weight (g/mol): 206.63
Molecular Formula: C11H7ClO2
InChi Key: YUCYPPIIPWVJMM-UHFFFAOYSA-N
SMILES: ClC(=O)C1=CC=C(O1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results