accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CSNOHN

CAS RN 5751-20-2

IUPAC Name: 2-(methylsulfanyl)-3,4-dihydropyrimidin-4-one
Synonyms: 2-(methylsulfanyl)-3H-pyrimidin-4-one
Molecular Weight (g/mol): 142.18
Molecular Formula: C5H6N2OS
InChi Key: UYHSQVMHSFXUOA-UHFFFAOYSA-N
SMILES: CSC1=NC=CC(=O)N1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results