accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 576-24-9

IUPAC Name: 2,3-dichlorophenol
Synonyms: 2,3-dichlorophenol
Molecular Weight (g/mol): 163
Molecular Formula: C6H4Cl2O
InChi Key: UMPSXRYVXUPCOS-UHFFFAOYSA-N
SMILES: OC1=CC=CC(Cl)=C1Cl
Molecular Weight (g/mol) 
  • (3)
  • (4)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results