accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 576-26-1

IUPAC Name: 2,6-dimethylphenol
Synonyms: 2,6-dimethylphenol
Molecular Weight (g/mol): 122.17
Molecular Formula: C8H10O
InChi Key: NXXYKOUNUYWIHA-UHFFFAOYSA-N
SMILES: CC1=CC=CC(C)=C1O
Molecular Weight (g/mol) 
  • (5)
  • (8)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (5)
Physical Form 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (1)
  • (5)
  • (8)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (5)
  • (5)
  • (1)
  • (1)