accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BrHOFBr

CAS RN 576-86-3

IUPAC Name: 2,4-dibromo-6-fluorophenol
Synonyms: 2,4-dibromo-6-fluorophenol
Molecular Weight (g/mol): 269.9
Molecular Formula: C6H3Br2FO
InChi Key: XJDJZAUITWTZAS-UHFFFAOYSA-N
SMILES: OC1=C(Br)C=C(Br)C=C1F
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results