accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 5761-97-7

CH3PClO

CAS RN 5761-97-7

IUPAC Name: methyl(phenyl)phosphinoyl chloride
Synonyms: methyl(phenyl)phosphinoyl chloride
Molecular Weight (g/mol): 174.56
Molecular Formula: C7H8ClOP
InChi Key: BTMQENOUJJKBGT-UHFFFAOYNA-N
SMILES: CP(Cl)(=O)C1=CC=CC=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)