CAS RN 5766-79-0
CAS RN 5766-79-0
IUPAC Name:
2-phenyl-2-(piperidin-1-yl)acetonitrile
Synonyms:
2-phenyl-2-(piperidin-1-yl)acetonitrile
Molecular Weight (g/mol):
200.29
Molecular Formula:
C13H16N2
InChi Key:
IXQJQHORYHYKBD-UHFFFAOYNA-N
SMILES:
N#CC(N1CCCCC1)C1=CC=CC=C1