accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OOOH

CAS RN 57668-34-5

IUPAC Name: 3-[3-(benzyloxy)phenyl]propanoic acid
Synonyms: 3-[3-(benzyloxy)phenyl]propanoic acid
Molecular Weight (g/mol): 256.3
Molecular Formula: C16H16O3
InChi Key: HVXOYUOKPLHWNU-UHFFFAOYSA-N
SMILES: OC(=O)CCC1=CC(OCC2=CC=CC=C2)=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results