accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 5768-39-8

IUPAC Name: 2H-1,3-benzodioxole-4-carboxylic acid
Synonyms: 2H-1,3-benzodioxole-4-carboxylic acid
Molecular Weight (g/mol): 166.13
Molecular Formula: C8H6O4
InChi Key: DBUAYOWCIUQXQW-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=CC2=C1OCO2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results