accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OOSH2NOO

CAS RN 57683-71-3

IUPAC Name: methyl 2-sulfamoylbenzoate
Synonyms: methyl 2-sulfamoylbenzoate
Molecular Weight (g/mol): 215.22
Molecular Formula: C8H9NO4S
InChi Key: VSOOBQALJVLTBH-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=CC=C1S(N)(=O)=O
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results