accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 57688-17-2

ONH2FFFCl

CAS RN 57688-17-2

IUPAC Name: 4-(4-chlorophenoxy)-3-(trifluoromethyl)aniline
Synonyms: 4-(4-chlorophenoxy)-3-(trifluoromethyl)aniline
Molecular Weight (g/mol): 287.67
Molecular Formula: C13H9ClF3NO
InChi Key: LEIVFIRDFAJXDA-UHFFFAOYSA-N
SMILES: NC1=CC(=C(OC2=CC=C(Cl)C=C2)C=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results