accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3H3CCH3NH2S

CAS RN 57774-77-3

IUPAC Name: 4-tert-butylbenzene-1-carbothioamide
Synonyms: 4-tert-butylbenzenecarbothioamide
Molecular Weight (g/mol): 193.31
Molecular Formula: C11H15NS
InChi Key: HZODWYBXBKXJLB-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC=C(C=C1)C(N)=S
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)