CAS RN 5779-93-1
CAS RN 5779-93-1
IUPAC Name:
2,3-dimethylbenzaldehyde
Synonyms:
2,3-dimethylbenzaldehyde
Molecular Weight (g/mol):
134.18
Molecular Formula:
C9H10O
InChi Key:
UIFVCPMLQXKEEU-UHFFFAOYSA-N
SMILES:
CC1=CC=CC(C=O)=C1C
1
–
1
of
1
results
1
2,3-Dimethylbenzaldehyde, 97%
CAS: 5779-93-1 Molecular Formula: C9H10O Molecular Weight (g/mol): 134.178 MDL Number: MFCD00798004 InChI Key: UIFVCPMLQXKEEU-UHFFFAOYSA-N Synonym: benzaldehyde, dimethyl,hemellitaldehyde,dimethylbenzaldehyde,o-xylene-3-carboxaldehyde,dimethylformylbenzene,xylene-formaldehyde,pubchem3080,acmc-1ake8,2,3-dimethyl-benzaldehyde,2,3-dimethyl benzaldehyde PubChem CID: 34224 IUPAC Name: 2,3-dimethylbenzaldehyde SMILES: CC1=CC=CC(=C1C)C=O