CAS RN 578-39-2
IUPAC Name:
4-hydroxy-2-methylbenzoic acid
Synonyms:
4-hydroxy-2-methylbenzoic acid
Molecular Weight (g/mol):
152.15
Molecular Formula:
C8H8O3
InChi Key:
BBMFSGOFUHEVNP-UHFFFAOYSA-N
SMILES:
CC1=CC(O)=CC=C1C(O)=O
1
–
1
of
1
results
1
4-Hydroxy-2-methylbenzoic acid, 98%, Thermo Scientific Chemicals
CAS: 578-39-2 Molecular Formula: C8H8O3 Molecular Weight (g/mol): 152.149 MDL Number: MFCD02182261 InChI Key: BBMFSGOFUHEVNP-UHFFFAOYSA-N Synonym: 4-hydroxy-o-toluic acid,4,2-cresotic acid,2-methyl-4-hydroxybenzoic acid,4-hydroxy-2-methyl-benzoic acid,benzoic acid, 4-hydroxy-2-methyl,2-methyl-4-hydroxybenzoicacid,acmc-209m0q,methyl p-hydroxybenzoic acid,ksc494s9j,4-hydroxy-2-methylbenzoicacid PubChem CID: 68475 IUPAC Name: 4-hydroxy-2-methylbenzoic acid SMILES: CC1=C(C=CC(=C1)O)C(=O)O