accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 5780-07-4

H3COOOO

CAS RN 5780-07-4

IUPAC Name: 7-methoxy-2H-1,3-benzodioxole-5-carbaldehyde
Synonyms: 7-methoxy-2H-1,3-benzodioxole-5-carbaldehyde
Molecular Weight (g/mol): 180.16
Molecular Formula: C9H8O4
InChi Key: LOFRBHZYZCIOJO-UHFFFAOYSA-N
SMILES: COC1=CC(C=O)=CC2=C1OCO2
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results