accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 57835-92-4

IUPAC Name: 4-nitropyrene
Synonyms: 4-nitropyrene
Molecular Weight (g/mol): 247.25
Molecular Formula: C16H9NO2
InChi Key: UISKIUIWPSPSAV-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC2=CC=CC3=CC=C4C=CC=C1C4=C23
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)