accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 5784-45-2

IUPAC Name: 1-chlorophthalazine
Synonyms: 1-chlorophthalazine
Molecular Weight (g/mol): 164.59
Molecular Formula: C8H5ClN2
InChi Key: UCOVESIAFFGEOR-UHFFFAOYSA-N
SMILES: ClC1=C2C=CC=CC2=CN=N1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results