accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OOHNH2N

CAS RN 5785-06-8

IUPAC Name: 3-methoxybenzohydrazide
Synonyms: benzoylhydrazine, M-methoxy
Molecular Weight (g/mol): 166.18
Molecular Formula: C8H10N2O2
InChi Key: VMZSDAQEWPNOIB-UHFFFAOYSA-N
SMILES: COC1=CC=CC(=C1)C(=O)NN
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results