accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 5786-21-0

IUPAC Name: 6-chloro-10-(4-methylpiperazin-1-yl)-2,9-diazatricyclo[9.4.0.0³,⁸]pentadeca-1,3,5,7,10,12,14-heptaene
Synonyms: clozaril iprox asaleptin clozapine (substance) leponex lepotex clozapine (product) fazaclo odt fazaclo clozapine preparation
Molecular Weight (g/mol): 326.83
Molecular Formula: C18H19ClN4
InChi Key: ZUXABONWMNSFBN-UHFFFAOYSA-N
SMILES: CN1CCN(CC1)C1=C2C=CC=CC2=NC2=CC=C(Cl)C=C2N1
Molecular Weight (g/mol) 
  • (10)
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Industry Type 
  • (1)
  • (1)
  • (4)
Certifications/Compliance 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (3)
  • (10)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)