accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COO

CAS RN 58-27-5

IUPAC Name: 2-methyl-1,4-dihydronaphthalene-1,4-dione
Synonyms: menadione kativ-G kolklot menaphthone menaphthene 甲萘醌 menadione (substance) memodol vitamin k>3< (substance) menaphthone preparation
Molecular Weight (g/mol): 172.18
Molecular Formula: C11H8O2
InChi Key: MJVAVZPDRWSRRC-UHFFFAOYSA-N
SMILES: CC1=CC(=O)C2=CC=CC=C2C1=O
Molecular Weight (g/mol) 
  • (4)
  • (6)
Percent Purity 
  • (6)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (3)
  • (1)
  • (4)
  • (6)
  • (6)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (6)