accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HClH3CNHN

CAS RN 58-28-6

IUPAC Name: hydrogen (3-{2-azatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,11,13-hexaen-2-yl}propyl)(methyl)amine chloride
Synonyms: desipramine hydrochloride desmethylimipramine chloride desmethylimipramine monohydrochloride desimipramine, hydrochloride desipramine hcl demethylimipramine hydrochloride desmethylimipramine hydrochloride dmi hydrochloride imipraminedemethyl hydrochloride desipramine monohydrochloride
Molecular Weight (g/mol): 302.85
Molecular Formula: C18H23ClN2
InChi Key: XAEWZDYWZHIUCT-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CNCCCN1C2=CC=CC=C2CCC2=CC=CC=C12
Industry Type 
  • (2)
  • (4)
Certifications/Compliance 
  • (2)
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (4)
  • (2)
  • (1)
  • (1)
  • (3)
  • (2)
  • (4)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)