accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CCH3CH3CH3H3CH3COCH3OCH3CH3O

CAS RN 58-95-7

IUPAC Name: 2,5,7,8-tetramethyl-2-(4,8,12-trimethyltridecyl)-3,4-dihydro-2H-1-benzopyran-6-yl acetate
Synonyms: tocopheryl acetate
Molecular Weight (g/mol): 472.75
Molecular Formula: C31H52O3
InChi Key: ZAKOWWREFLAJOT-UHFFFAOYNA-N
SMILES: CC(C)CCCC(C)CCCC(C)CCCC1(C)CCC2=C(C)C(OC(C)=O)=C(C)C(C)=C2O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results